Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO18B Monoclonal Antibody (1683CT270.39.50)
GREENER_CHOICE

Catalog No. PIMA524752
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524752 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524752 Supplier Invitrogen™ Supplier No. MA524752
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYO18B Monoclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene may regulate muscle-specific genes when in the nucleus and may influence intracellular trafficking when in the cytoplasm. The encoded protein functions as a homodimer and may interact with F actin. Mutations in this gene are associated with lung cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO18B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1683CT270.39.50
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MYO18B
Gene Accession No. Q8IUG5
Gene Alias DKFZp434C2422; DKFZp779C1668; FLJ41942; KFS4; MY18B; MYO18B; myosin 18B; myosin XVIIIB; Myosin-XVIIIb; unconventional myosin-XVIIIb
Gene Symbols MYO18B
Host Species Mouse
Immunogen human MYO18B recombinant protein.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84700
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.