Learn More
Invitrogen™ MYO18B Polyclonal Antibody

Description
The antibody detects endogenous levels of total MYO18B protein.
Specifications
Specifications
| Antigen | MYO18B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | MYO18B |
| Gene Accession No. | Q8IUG5 |
| Gene Alias | DKFZp434C2422; DKFZp779C1668; FLJ41942; KFS4; MY18B; MYO18B; myosin 18B; myosin XVIIIB; Myosin-XVIIIb; unconventional myosin-XVIIIb |
| Gene Symbols | MYO18B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.