Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563820 Supplier Invitrogen™ Supplier No. PA563820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EVVNKINRAN GKSTSRIFLL TNNNLLLADQ KSGQIKSEVP LVDVTKVSMS SQNDGFFAVH LKEGSEAASK GDFLFSSDHL IEMATKLYRT TLSQTKQKLN IEISDEFLVQ FR Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO1B
Gene Accession No. O43795
Gene Alias AA406997; MIH-L; MMIa; MMI-alpha; molecular motor; MYH-1c; Myo1a; MYO1B; MYO1B variant protein; myosin heavy chain myr 1; Myosin I alpha; Myosin IA (Myosin I alpha) (MMI-alpha) (MMIa) (MIH-L); myosin Ib; myosin-I alpha; myosin-Ib; myr1; Unconventional myosin-Ib
Gene Symbols MYO1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO1B. Recombinant protein control fragment (Product #RP-103368).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4430
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.