Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142637
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142637 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142637 Supplier Thermo Scientific Supplier No. PA5142637
Add to Cart
Add to Cart
This item is not returnable. View return policy

MYO1B Polyclonal Antibody

MYO1B Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MYO1B
Gene Accession No. O43795, P46735
Gene Alias AA406997; MIH-L; MMIa; MMI-alpha; molecular motor; MYH-1c; Myo1a; MYO1B; MYO1B variant protein; myosin heavy chain myr 1; Myosin I alpha; Myosin IA (Myosin I alpha) (MMI-alpha) (MMIa) (MIH-L); myosin Ib; myosin-I alpha; myosin-Ib; myr1; Unconventional myosin-Ib
Gene Symbols MYO1B
Host Species Goat
Immunogen Peptide with sequence C-RTVEAKQEKVSTT.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17912, 4430
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.