Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1B Recombinant Rabbit Monoclonal Antibody (JE65-01)
GREENER_CHOICE

Catalog No. FEMA550668
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550668 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550668 Supplier Invitrogen™ Supplier No. MA550668
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: A431 cell lysate, NIH/3T3 cell lysate, human lymph nodes tissue, human testis tissue, human colon carcinoma tissue. Predicted band size: 132 kDa Subcellular Location: Actin cytoskeleton, actin filament, myosin complex, early endosome, endosome membrane, extracellular exosome, plasma membrane, apical part of cell, brush border, cell periphery, cytoplasm, filopodium, microvillus, perinuclear region of cytoplasm, vesicle.

Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE65-01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MYO1B
Gene Accession No. O43795, P46735
Gene Alias AA406997; MIH-L; MMIa; MMI-alpha; molecular motor; MYH-1c; Myo1a; MYO1B; MYO1B variant protein; myosin heavy chain myr 1; Myosin I alpha; Myosin IA (Myosin I alpha) (MMI-alpha) (MMIa) (MIH-L); myosin Ib; myosin-I alpha; myosin-Ib; myr1; Unconventional myosin-Ib
Gene Symbols MYO1B
Host Species Rabbit
Immunogen Recombinant protein within human MYO1B aa 916-1136.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17912, 4430
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.