Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564905
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564905 Supplier Invitrogen™ Supplier No. PA564905
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEEETLRRLGL ERNPQSYLYL VKGQCAKVSS INDKSDWKVV RKALTVIDFT EDEVEDLLSI VASVLHLGNI HFAANEESNA QVTTENQLKY LTRLLSVEGS TLREALTHRK IIAKGEELLS PLNLEQAAYA RDALAKAVYS RTFTWLV Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.

MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1C
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO1C
Gene Accession No. O00159
Gene Alias C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic
Gene Symbols MYO1C
Host Species Rabbit
Immunogen Recombinant protein fragment of Human MYO1C (Product #RP-100988)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4641
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.