Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1C Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA593297

Catalog No. PIPA593297


Only null left
Add to Cart

Description

Description

Immunogen sequence: ENAFFLDHVR TSFLLNLRRQ LPQNVLDTSW PTPPPALREA SELLRELCIK NMVWKYCRSI SPEWKQQLQQ KAVASEIFKG KKDNYPQSVP RLFISTRLGT DEISPRVLQA LGSEPIQYAV PVVKYDRKGY KPRSRQLLLT PNAVVIVEDA KVKQRIDYAN LTGISVSSLS DSLFVLHVQR ADNKQKGDVV LQSDHVIETL TKTALSANRV NSININQGSI TFAGGPGRDG TIDFTPGSEL LITKAKNGHL AVVAPRLNSR; Positive Samples: HT-29, SKOV3, Mouse lung, Rat lung; Cellular Location: Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Cytoplasmic vesicle, Nucleus, Nucleus, Peripheral membrane protein, nuclear pore complex, nucleolus, nucleoplasm, ruffle, stereocilium membrane.

MYO1C is a member of the unconventional myosin protein family, which are actin-based molecular motors. The protein is found in the cytoplasm, and one isoform with a unique N-terminus is also found in the nucleus. The nuclear isoform associates with RNA polymerase I and II and functions in transcription initiation. The mouse ortholog of this protein also functions in intracellular vesicle transport to the plasma membrane.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MYO1C
Polyclonal
Unconjugated
MYO1C
C80397; FLJ23903; mm1beta; MMIb; MMI-beta; myo1c; myo1ca; MYO1E; myosin 1c, paralog a; myosin heavy chain myr 2; myosin I; Myosin I beta; myosin IB; myosin IC; myosin Ic, paralog a; myosin IC-like; myosin-I beta; myosin-Ibeta; myosin-Ic; Myr2; NMI; nuclear myosin I; nuclear myosin I beta; si:ch211-69i14.4; unconventional myosin Myr2 I heavy chain; unconventional myosin-Ic
Rabbit
Affinity Chromatography
RUO
17913, 4641, 65261
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.95 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00159, Q63355, Q9WTI7
MYO1C
Recombinant fusion protein containing a sequence corresponding to amino acids 769-1028 of human MYO1C (NP_2036933).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.