Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119295
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119295 200 μL
PIPA5121750 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119295 Supplier Invitrogen™ Supplier No. PA5119295
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MYO1D Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Unconventional myosin that functions as actin-based motor protein with ATPase activity. Plays a role in endosomal protein trafficking, and especially in the transfer of cargo proteins from early to recycling endosomes. Required for normal planar cell polarity in ciliated tracheal cells, for normal rotational polarity of cilia, and for coordinated, unidirectional ciliary movement in the trachea. Required for normal, polarized cilia organization in brain ependymal epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1D
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MYO1D
Gene Accession No. Q63357
Gene Alias 9930104H07Rik; AW544947; D11Ertd9e; KIAA0727; Myo1c; MYO1D; myosin 1C; myosin heavy chain myr 4; myosin IC; myosin ID; myosin1C; myosin-1d; myosin-I gamma; myosin-Id; Myr2; Myr4; PPP1R108; protein phosphatase 1, regulatory subunit 108; Unconventional myosin from rat 4 for myosin I heavy chain; unconventional myosin-Id
Gene Symbols MYO1D
Host Species Rabbit
Immunogen Recombinant protein Myosin ID. The antigen corresponds to amino acid range 512-788 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25485
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.