Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120743
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120743 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120743 Supplier Invitrogen™ Supplier No. PA5120743
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SKOV3, Mouse lung, Mouse brain. Immunogen sequence: HINNYLLEKS RVIVQQPGER SFHSFYQLLQ GGSEQMLRSL HLQKSLSSYN YIHVGAQLKS SINDAAEFRV VADAMKVIGF KPEEIQTVYK ILAAILHLGN LKFVVDGDTP LIENGKVVSI IAELLSTKTD MVEKALLYRT VATGRDIIDK QHTEQEASYG RDAFAKAIYE RLFCWIVTRI NDIIEVKNYD TTIHGKNTVI G.

Unconventional myosin that functions as actin-based motor protein with ATPase activity. Plays a role in endosomal protein trafficking, and especially in the transfer of cargo proteins from early to recycling endosomes. Required for normal planar cell polarity in ciliated tracheal cells, for normal rotational polarity of cilia, and for coordinated, unidirectional ciliary movement in the trachea. Required for normal, polarized cilia organization in brain ependymal epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1D
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.276 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene MYO1D
Gene Accession No. O94832, Q5SYD0
Gene Alias 9930104H07Rik; AW544947; D11Ertd9e; KIAA0727; Myo1c; MYO1D; myosin 1C; myosin heavy chain myr 4; myosin IC; myosin ID; myosin1C; myosin-1d; myosin-I gamma; myosin-Id; Myr2; Myr4; PPP1R108; protein phosphatase 1, regulatory subunit 108; Unconventional myosin from rat 4 for myosin I heavy chain; unconventional myosin-Id
Gene Symbols MYO1D
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-380 of human MYO1D (NP_001290209.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338367, 4642
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.