Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554913 Supplier Invitrogen™ Supplier No. PA554913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIRNQYVPYP HAPGSQRSNQ KSLYTSMARP PLPRQQSTSS DRVSQTPESL DFLKVPDQGA AGVRRQTTSR PPPAGGRPKP QPKPKPQVPQ CKALYAYDAQ DTDELSFNAN DIIDIIKEDP SGWWTGRLRG KQG Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1E
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO1E
Gene Accession No. Q12965
Gene Alias 2310020N23Rik; 9130023P14Rik; AA407778; FSGS6; HuncM-IC; LOW QUALITY PROTEIN: unconventional myosin-Ie; molecular motor; MYO1C; Myo1e; MYO1E variant protein; myosin heavy chain myr 3; myosin IE; myosin-1e; myosin-IC; myosin-Ie; myosin-Ie; unconventional myosin-Ie; myr 3; Myr3; MYR5; unconventional myosin 1E; Unconventional myosin from rat 5; unconventional myosin-Ie
Gene Symbols MYO1E
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO1E. Recombinant protein control fragment (Product #RP-101032).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4643
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.