Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588931 Supplier Invitrogen™ Supplier No. PA588931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KEVLKRKIEI ERILSVSLST MQDDIFILHE QEYDSLLESV FKTEFLSLLA KRYEEKTQKQ LPLKFSNTLE LKLKKENWGP WSAGGSRQVQ FHQGFGDLAV LKPSNKVLQV SIGPGLPKNS RPTRRNTTQN TGYSSGTQNA NYPVRAAPPP PGYHQNGVIR NQYVPYPHAP GSQRSNQKSL YTSMARPPLP RQQSTSSDRV SQTPESLDFL KVPDQGAAGV RRQTTSRPPP AGGRPKPQPK PKPQVPQCKA LYAYDAQDTD ELSFNANDII DIIKEDPSGW WTGRLRGKQG LFPNNYVTKI; Positive Samples: HeLa; Cellular Location: Cell junction, Cytoplasm, Cytoplasmic vesicle, clathrin-coated vesicle, cytoskeleton.

Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1E
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MYO1E
Gene Accession No. Q12965
Gene Alias 2310020N23Rik; 9130023P14Rik; AA407778; FSGS6; HuncM-IC; LOW QUALITY PROTEIN: unconventional myosin-Ie; molecular motor; MYO1C; Myo1e; MYO1E variant protein; myosin heavy chain myr 3; myosin IE; myosin-1e; myosin-IC; myosin-Ie; myosin-Ie; unconventional myosin-Ie; myr 3; Myr3; MYR5; unconventional myosin 1E; Unconventional myosin from rat 5; unconventional myosin-Ie
Gene Symbols MYO1E
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 809-1108 of human MYO1E (NP_0049892).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.