Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYO1F Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546728
Description
Peptide sequence: CIKPNETKRP RDWEENRVKH QVEYLGLKEN IRVRRAGFAY RRQFAKFLQR.
Myosins are actin-based motor molecules with ATPase activity. Unconventional myosins serve in intracellular movements. Their highly divergent tails are presumed to bind to membranous compartments, which would be moved relative to actin filaments.
Specifications
MYO1F | |
Polyclonal | |
Unconjugated | |
MYO1F | |
C330006B10Rik; MYO1F; myosin IF; myosin-1F-like protein; myosin-ID; myosin-Ie; myosin-If; unconventional myosin-If | |
Rabbit | |
Affinity chromatography | |
RUO | |
4542 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O00160 | |
MYO1F | |
Synthetic peptide directed towards the C-terminal of human MYO1F (aa 584-633). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction