Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYO1H Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558825
Description
Immunogen sequence: VSTSNLSDGI LVIHVSPEDS KQKGDAVLQC GHVFEAVTKL VMLVKKENIV NVVQGSLQFF ISPGKEGTIV FDTGPEEQVY KNKNGQLTVV SG Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 72%.

Specifications
MYO1H | |
Polyclonal | |
Unconjugated | |
MYO1H | |
4631401O15Rik; MYO1H; myosin 1H; myosin IH; myosin-1H; myosin-Ih; unconventional myosin-Ih | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283446 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N1T3 | |
MYO1H | |
Recombinant protein corresponding to Human MYO1H. Recombinant protein control fragment (Product #RP-97851). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction