Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO1H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537996
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537996 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537996 Supplier Invitrogen™ Supplier No. PA537996
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with human, rat, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO1H
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MYO1H
Gene Accession No. Q8N1T3, Q9D6A1
Gene Alias 4631401O15Rik; MYO1H; myosin 1H; myosin IH; myosin-1H; myosin-Ih; unconventional myosin-Ih
Gene Symbols MYO1H
Host Species Goat
Immunogen Peptide with sequence C-EDQVYKDKNGQLR, from the C-terminus of Myo1H.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231646, 283446
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.