Learn More
Promega MYO3 beta Kinase Enzyme System, 1mg
Catalog No. PRV6452 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6452 1 mg
1 options
Catalog No. PRV6452 Supplier Promega Supplier No. V6452
Add to Cart
Add to Cart
Request a Quote
Recombinant human MYO3 beta (1-326) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MYO3 beta is a member of the Class III myosins that are actin-dependent motor proteins containing an amino-terminal kinase domain.
  • Easily Screen and Profile MYO3β Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C