Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO3A Monoclonal Antibody (8D12)
GREENER_CHOICE

Catalog No. PIMA524580
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524580 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524580 Supplier Invitrogen™ Supplier No. MA524580
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: HEEINNIKKK DNKDSKATSE REACGLAIFS KQISKLSEEY FILQKKLNEM ILSQQLKSLY LGVSHHKPIN RRVSSQQCLS GVCKGEEPKI L.

MYO3A (MYO3 alpha) serine/threonine kinase belongs to the myosin superfamily where they are categorized into conventional myosins (class II) and unconventional myosins (classes I and III through XV) based on their variable C-terminal cargo-binding domains. Class III myosins, such as MYO3A, have a kinase domain N-terminal to the conserved N-terminal motor domains and are expressed in photoreceptors. MYO3A plays an important role in hearing in humans and mutations in the MYO3A have been shown to cause nonsyndromic progressive hearing loss. MYO3A is highly expressed in retina and cochlea.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO3A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8D12
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MYO3A
Gene Accession No. Q8NEV4
Gene Alias DFNB30; MYO3A; myosin IIIA; myosin-IIIa
Gene Symbols MYO3A
Host Species Mouse
Immunogen MYO3A (NP_059129, 1400 a.a. ~ 1490 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53904
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.