Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140174 Supplier Invitrogen™ Supplier No. PA5140174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: KRRPRKDSQG KLLDLEDFYY KEFLPSRSGP KEHSPSLRER RPQQELQNQC IKANERCWAA ESPEKEEERE PAANPYDFR Highest antigen sequence indentity to the following orthologs: Rat - 62%, Mouse - 61%.

MYO3A (MYO3 alpha) serine/threonine kinase belongs to the myosin superfamily where they are categorized into conventional myosins (class II) and unconventional myosins (classes I and III through XV) based on their variable C-terminal cargo-binding domains. Class III myosins, such as MYO3A, have a kinase domain N-terminal to the conserved N-terminal motor domains and are expressed in photoreceptors. MYO3A plays an important role in hearing in humans and mutations in the MYO3A have been shown to cause nonsyndromic progressive hearing loss. MYO3A is highly expressed in retina and cochlea.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO3A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO3A
Gene Accession No. Q8NEV4
Gene Alias DFNB30; MYO3A; myosin IIIA; myosin-IIIa
Gene Symbols MYO3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO3A. Recombinant protein control fragment (Product #RP-106356).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53904
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.