Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYO3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MYO3B |
---|---|
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MYO3B Polyclonal specifically detects MYO3B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MYO3B | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q8WXR4 | |
140469 | |
This antibody was developed against a recombinant protein corresponding to amino acids: FISQCLIKDFERRPSVTHLLDHPFIKGVHGKVLFLQKQLAKVLQDQKHQNPVAKTRHERMHTRRPYHVEDAEKYCLEDDLVNLEVLDEDTIIHQLQKRYADLLIY | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
EC 2.7.11, EC 2.7.11.1, myosin IIIB, myosin-IIIb | |
MYO3B | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title