Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519366
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519366 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519366 Supplier Invitrogen™ Supplier No. PA519366
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

This gene is one of three myosin V heavy-chain genes, belonging to the myosin gene superfamily. Myosin V is a class of actin-based motor proteins involved in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. The protein encoded by this gene is abundant in melanocytes and nerve cells. Mutations in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and neuroectodermal melanolysosomal disease, or Elejalde disease. Multiple alternatively spliced transcript variants encoding different isoforms have been reported, but the full-length nature of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO5A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MYO5A
Gene Accession No. Q99104, Q9Y4I1
Gene Alias 9630007J19Rik; AI413174; AI661011; D; d-120J; Dbv; Dilute; dilute lethal-20J protein; dilute myosin heavy chain, non-muscle; dilute-opisthotonus; Dop; flail; flailer; flr; GS1; MVa; MYH12; MYO5; Myo5a; myosin 5a; Myosin heavy chain 12; Myosin heavy chain p190; myosin I heavy chain isoform; myosin V; myosin VA; myosin VA (heavy chain 12, myosin); myosin VA (heavy chain 12, myoxin); myosin VA (heavy polypeptide 12, myoxin); myosin, heavy polypeptide kinase; myosin-12; Myosin-V; myosin-Va; MyoVA; myoxin; MYR12; OTTMUSP00000017756; OTTMUSP00000019176; OTTMUSP00000046608; p190 myosin heavy chain; Sev-1; unconventional myosin-Va
Gene Symbols MYO5A
Host Species Goat
Immunogen Synthetic peptide sequence (ETKQLELDLN) corresponding to the internal amino acids of MYO5A (aa 1063-1072).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17918, 4644
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.