Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599065 Supplier Invitrogen™ Supplier No. PA599065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYO5B Polyclonal Antibody for Western Blot, IHC (P), ELISA

May be involved in vesicular trafficking via its association with the CART complex. The CART complex is necessary for efficient transferrin receptor recycling but not for EGFR degradation. Required in a complex with RAB11A and RAB11FIP2 for the transport of NPC1L1 to the plasma membrane. Together with RAB11A participates in CFTR trafficking to the plasma membrane and TF (transferrin) recycling in nonpolarized cells. Together with RAB11A and RAB8A participates in epithelial cell polarization. Together with RAB25 regulates transcytosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO5B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYO5B
Gene Accession No. Q9ULV0
Gene Alias AI661750; KIAA1119; mKIAA1119; MYO5B; MYO5B variant protein; myosin 5B; myosin heavy chain myr 6; Myosin of the dilute-myosin-V family; myosin VB; myosin-Vb; Myr6; Unconventional myosin-Vb
Gene Symbols MYO5B
Host Species Rabbit
Immunogen Recombinant Human Unconventional myosin-Vb protein (1440-1690AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4645
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.