Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568237 Supplier Invitrogen™ Supplier No. PA568237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human, Mouse, and Rat samples.

May be involved in transferrin trafficking. Likely to power actin-based membrane trafficking in many physiologically crucial tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO5C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYO5C
Gene Accession No. Q9NQX4
Gene Alias 9130003O20Rik; BB085560; C330029I24; MYO5C; myosin 5C; myosin VC; myosin-Vc; unconventional myosin-Vc
Gene Symbols MYO5C
Host Species Rabbit
Immunogen Synthesized peptide derived from human MYO5C.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208943, 315820, 55930
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.