Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO6/Myosin VI Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1216 Supplier Bioss Supplier No. BS11264R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein involved intracellular vesicle and organelle transport, especially in the hair cell of the inner ear. Mutations in this gene have been found in patients with non-syndromic autosomal dominant and recessive hearing loss.

Specifications

Antigen MYO6/Myosin VI
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MYO6
Gene Accession No. Q9UM54
Gene Alias BC029719; DFNA22; DFNB37; KIAA0389; MYO6; myosin VI; myosin-VI; RGD1560646; Snell′s waltzer; sv; tailchaser; Tlc; Unconventional myosin-6; Unconventional myosin-VI
Gene Symbols MYO6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Myosin VI.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4646
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.