Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYO7A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733052 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3052 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3052 Supplier Proteintech Group Inc Supplier No. 207201AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody is specific to MYO7A.

Myosin VIIa is a member of the myosin superfamily of actin-based motor proteins. Defects in the myosin VIIa gene are responsible for hearing impairment in shaker-1 (sh1) mice and causes Usher syndrome IB in humans. Usher syndrome associates congenital deafness, vestibular dysfunction, and retinitis pigmentosa and is the most common form of combined deafness and blindness. Structural features of myosin VIIa protein include an ATP binding N-terminal motor domain, a central region which possess five light-chain binding (IQ) motifs, and a C-terminal domain with three myosin tail homology (MyTH4) and talin-like homology regions.

Specifications

Antigen MYO7A
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MYO7A
Gene Accession No. P97479, Q13402
Gene Alias DFNA11, DFNB2, MYO7A, myosin VIIA, MYOVIIA, MYU7A, NSRD2, USH1B
Gene Symbols Myo7a
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17921, 266714, 4647
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.