Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYO9A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559055
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559055 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559055 Supplier Invitrogen™ Supplier No. PA559055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSQRFSSVDE QAKLHKTMSQ GEITKLAVRQ KASDSDIRPQ RAKMRFWAKG KQGEKKTTRV KPTTQSEVSP LFAGTDVIPA HQFPDELAA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

This gene encodes a member of the myosin superfamily. The protein represents an unconventional myosin; it should not be confused with the conventional non-muscle myosin-9 (MYH9). Unconventional myosins contain the basic domains of conventional myosins and are further distinguished from class members by their tail domains. They function as actin-based molecular motors. Mutations in this gene have been associated with Bardet-Biedl Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYO9A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MYO9A
Gene Accession No. B2RTY4
Gene Alias FLJ11061; FLJ13244; MGC71859; MYO9A; myosin IXA; myosin-IXa; MYR7; Unconventional myosin-9a; unconventional myosin-IXa
Gene Symbols MYO9A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYO9A. Recombinant protein control fragment (Product #RP-98036).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.