Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYO9B Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298029H
Description
MYO9B Polyclonal antibody specifically detects MYO9B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
MYO9B | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
CELIAC4, myosin IXB, myosin-IXb, MYR5Unconventional myosin-9b, unconventional myosin IXb | |
Produced in rabbits immunized with E. coli-derived Human MYO9B fragment. | |
0.1 mL | |
Signal Transduction | |
4650 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
HRP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction