Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYO9B Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10041092
Description
MYO9B Polyclonal specifically detects MYO9B in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.Specifications
MYO9B | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CELIAC4, myosin IXB, myosin-IXb, MYR5Unconventional myosin-9b, unconventional myosin IXb | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_004136.2; NP_001123537.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 2 - 6 ug/ml, Peptide ELISA Detection limit 1:2000 | |
NP_004136.2 | |
MYO9B | |
Peptide with sequence C-KNDFEPVKQSKAQ corresponding to internal region according to NP_004136.2, NP_001123537.1. | |
0.1 mg | |
Signal Transduction | |
4650 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction