Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYO9B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559838
Description
Immunogen sequence: TDGERSAKKP AVQKKKPGDA SSLPDAGLSP GSQVDSKSTF KRLFLHKTKD KKYSLEGAEE LENAVSGHVV LEATTM Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.
This gene encodes a member of the myosin family of actin-based molecular motor heavy chain proteins. The protein has four IQ motifs located in the neck domain that bind calmodulin, which serves as a light chain. The protein complex has a single-headed structure and exhibits processive movement on actin filaments toward the minus-end. The protein also has rho-GTPase activity. Polymorphisms in this gene are associated with celiac disease and ulcerative colitis susceptibility. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
MYO9B | |
Polyclonal | |
Unconjugated | |
MYO9B | |
celiac disease 4; CELIAC4; MYO9B; myosin; myosin IXb; myosin-IXb; myr 5; MYR5; Unconventional myosin from rat 3; unconventional myosin IXb; unconventional myosin-9b; Unconventional myosin-IXb | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4650 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13459 | |
MYO9B | |
Recombinant protein corresponding to Human MYO9B. Recombinant protein control fragment (Product #RP-98904). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction