Learn More
Invitrogen™ MYOC Polyclonal Antibody

Description
Immunogen sequence: CGELVWVGEP LTLRTAETIT GKYGVWMRDP KPTYPYTQET TWRIDTVGTD VRQVFEYDLI SQFMQGYPSK VHILPRPLES TGAVVYSGSL YFQGAESRTV IRYELNTETV KAEKEIPGAG YHGQFPYSWG GYTDIDLAVD EAGLWVIYST DEAKGAIVLS KLNPENLELE QTWETNIRKQ SVANAFIICG TLYTVSSYTS ADATVNFAYD TGTGISKTLT IPFKNRYKYS SMIDYNPLEK KLFAWDNLNM VTYDIKLSKM; Positive Samples: Raji, U-937, BT-474, Mouse brain; Cellular Location: Cell projection, Cytoplasmic vesicle, Endoplasmic reticulum, Golgi apparatus, Mitochondrion, Mitochondrion inner membrane, Mitochondrion intermembrane space, Mitochondrion outer membrane, Rough endoplasmic reticulum, cilium, exosome, extracellular matrix, extracellular space.
Specifications
Specifications
| Antigen | MYOC |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.46 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MYOC |
| Gene Accession No. | O70624, Q99972 |
| Gene Alias | AI957332; GLC1A; GPOA; JOAG; JOAG1; juvenile-onset open-angle glaucoma 1; mutated trabecular meshwork-induced glucocorticoid response protein; MYOC; Myocilin; Myocilin 20 kDa N-terminal fragment; Myocilin 35 kDa N-terminal fragment; Myocilin 55 kDa subunit; myocilin trabecular meshwork inducible glucocorticoid response; myocilin trabecular meshwork inducible glucocorticoid response protein; Myocilin, C-terminal fragment; Myocilin, N-terminal fragment; myocilin, trabecular meshwork inducible glucocorticoid response; TIGR; trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork-induced glucocorticoid response protein |
| Gene Symbols | MYOC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.