Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595700 Supplier Invitrogen™ Supplier No. PA595700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat heart; Cellular Location: Nucleus Immunogen sequence: SQVCTAQMAG LHSSDKVGPK FSIPSPTFSK SSSAISEVTQ PPSYEDAVKQ QMTRSQQMDE LLDVLIESGE MPADAREDHS CLQKVPKIPR SSRSPTAVLT K.

Smooth muscle cells (SM) and cardiac muscle cells-specific transcriptional factor which uses the canonical single or multiple CArG boxes DNA sequence. Acts as a cofactor of serum response factor (SRF) with the potential to modulate SRF-target genes. Plays a crucial role in cardiogenesis and differentiation of the smooth muscle cell lineage (myogenesis).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOCD
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Myocd
Gene Accession No. Q8IZQ8, Q8R5I7
Gene Alias Basic SAP coiled-coil transcription activator 2; Bsac2; BSAC2A; Mycd; Myocardin; Myocd; SRF cofactor protein; SRF co-factor protein (cardiac and smooth muscle); Srfcp; transcription factor myocardin
Gene Symbols Myocd
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 680-780 of human MYOCD (NP_7058321).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246297, 93649
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.