Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139907 Supplier Invitrogen™ Supplier No. PA5139907
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ANQGIIPPLK RPAEFHEQRK HLDSDKAKNS LKRKARNRCN SADLVNMHIL QASTAERSIP TAQM Highest antigen sequence indentity to the following orthologs: Rat - 75%, Mouse - 73%.

Smooth muscle cells (SM) and cardiac muscle cells-specific transcriptional factor which uses the canonical single or multiple CArG boxes DNA sequence. Acts as a cofactor of serum response factor (SRF) with the potential to modulate SRF-target genes. Plays a crucial role in cardiogenesis and differentiation of the smooth muscle cell lineage (myogenesis).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOCD
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Myocd
Gene Accession No. Q8IZQ8
Gene Alias Basic SAP coiled-coil transcription activator 2; Bsac2; BSAC2A; Mycd; Myocardin; Myocd; SRF cofactor protein; SRF co-factor protein (cardiac and smooth muscle); Srfcp; transcription factor myocardin
Gene Symbols Myocd
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MYOCD. Recombinant protein control fragment (Product #RP-109261).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.