Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myocilin Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502207873
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7873 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7873 Supplier Fabgennix Inc Supplier No. MYO101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myocilin Polyclonal antibody specifically detects Myocilin in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot

Specifications

Antigen Myocilin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene MYOC
Gene Accession No. O70624, Q99972, Q9R1J4
Gene Alias AI957332; GLC1A; GPOA; JOAG; JOAG1; juvenile-onset open-angle glaucoma 1; mutated trabecular meshwork-induced glucocorticoid response protein; MYOC; Myocilin; Myocilin 20 kDa N-terminal fragment; Myocilin 35 kDa N-terminal fragment; Myocilin 55 kDa subunit; myocilin trabecular meshwork inducible glucocorticoid response; myocilin trabecular meshwork inducible glucocorticoid response protein; Myocilin, C-terminal fragment; Myocilin, N-terminal fragment; myocilin, trabecular meshwork inducible glucocorticoid response; TIGR; trabecular meshwork inducible glucocorticoid response protein; trabecular meshwork-induced glucocorticoid response protein
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on Myocilin protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17926, 457511, 4653, 81523
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.