Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyoD Antibody (rMYD712), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308505AF594
Description
MyoD Monoclonal antibody specifically detects MyoD in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
MyoD | |
Monoclonal | |
Alexa Fluor 594 | |
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
Recombinant full-length human MyoD protein (Uniprot: P15172) | |
100 μg | |
Cancer, Cell Cycle and Replication, Growth and Development | |
4654 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunohistochemistry (Paraffin), Protein Array | |
rMYD712 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction