Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MyoD Antibody (SPM427), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234772MFV450
Description
MyoD Monoclonal antibody specifically detects MyoD in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
MyoD | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken | |
Purified |
Flow Cytometry | |
SPM427 | |
Flow Cytometry, Flow (Intracellular) | |
bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM | |
Recombinant MyoD of Mouse origin with an epitope mapping to amino acids 180-189. (Uniprot: P15172) | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific | |
4654 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction