Learn More
MyoD Monoclonal Antibody (5F11), Active Motif™

Description
Specifications
Specifications
| Antigen | MyoD |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | 5F11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 |
| Gene | MYOD1 |
| Gene Accession No. | P10085, P15172 |
| Gene Alias | AI503393; BHLHC1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; Myoblast determination protein 1 homolog; MYOD; Myod1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; Myogenic factor 3; PUM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.