Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MYOD Recombinant Rabbit Monoclonal Antibody (RM369)
GREENER_CHOICE

Catalog No. PIMA533061
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533061 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533061 Supplier Invitrogen™ Supplier No. MA533061
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: reacts to human, mouse, and rat MyoD1 (Myoblast determination protein 1). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MYOD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone RM369
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide
Gene MYOD1
Gene Accession No. P10085, P15172, Q02346
Gene Alias AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product
Gene Symbols MYOD1
Host Species Rabbit
Immunogen A peptide corresponding to residues near the N-terminus of human MyoD1 (Myoblast determination protein 1).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17927, 337868, 4654
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.