Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyoD1 Ab-2 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS278P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS278P1 500 μL 200μg/mL
MS278P0 100 μL 200μg/mL
MS278P 1 mL 200μg/mL
MS278P1ABX 100 μL 1mg/mL
MS278PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS278P1 Supplier Epredia Supplier No. MS278P1

Deliver accurate, reproducible results with the Epredia™ MyoD1 Ab-2, Mouse Monoclonal Antibody.

Research Areas: Muscle Markers; MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occassionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm's tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing's sarcomas and alveolar soft part sarcomas.

Host Species: Mouse

Clone: 5.2F

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Chicken. Others not known.

Epitope: aa 3-56

Immunogen: Recombinant mouse MyoD1 protein

Molecular Weight: 45kDa

Positive Control: SW80 cells

Cellular Localization: Nuclear

Recommended for:

  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen MyoD1 Ab-2
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 5.2F
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant mouse MyoD1 protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.