Learn More
Description
Host Species: Mouse
Clone: 5.2F
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Chicken. Others not known.
Epitope: aa 3-56
Immunogen: Recombinant mouse MyoD1 protein
Molecular Weight: 45kDa
Positive Control: SW80 cells
Cellular Localization: Nuclear
Recommended for:
- Immunofluorescence
- Immunoprecipitation (Native verified)
- Electron Microscopy
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | MyoD1 Ab-2 |
| Applications | Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 5.2F |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant mouse MyoD1 protein |
| Quantity | 200 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.