Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyoD1 Rabbit anti-Human, Clone: ZR262, Cocktail, Zeta

Catalog No. 502216315
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-221-6315 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-221-6315 Supplier ZETA CORPORATION Supplier No. Z2576RP
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody Cocktail

This product is diluted and in a ready-to-use formulation. A recommended positive control tissue for this product is Rhabdomyosarcoma, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.

MyoD1 Recombinant Monoclonal antibody specifically detects MyoD1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MyoD1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZR262
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene MYOD1
Gene Accession No. P15172
Gene Alias AI503393; bHLHc1; Class C basic helix-loop-helix protein 1; CMD1; MYF3; Myf-3; Myoblast determination protein 1; myoblast determination protein 1 homolog; Myod; MYOD1; Myod-1; MYOD1 homolog; myogenic differentiation 1; myogenic factor 1; myogenic factor 3; PUM; unnamed protein product
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues within aa 1-100 of human MyoD1
Purification Method Protein A
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4654
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.