Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myoferlin Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232264B
Description
Myoferlin Polyclonal antibody specifically detects Myoferlin in Human samples. It is validated for Western BlotSpecifications
Myoferlin | |
Polyclonal | |
Western Blot | |
fer-1 (C.elegans)-like 3 (myoferlin), FER1L3, fer-1-like 3, myoferlin, fer-1-like 3, myoferlin (C. elegans), Fer-1-like protein 3, FLJ36571, FLJ90777, KIAA1207, myoferlin | |
The immunogen this antibody was made to, maps to a region between residue 700 to 750 of human Myoferlin using the numbering given in entry NP_038479.1 (GeneID 26509). | |
0.1 mL | |
Cardiovascular Biology, Cell Biology | |
26509 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction