Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myofibroblast Marker Antibody (PR 2D3), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308768AF488
Description
Myofibroblast Marker Monoclonal antibody specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Myofibroblast Marker | |
Monoclonal | |
Alexa Fluor 488 | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
PR 2D3 | |
50mM Sodium Borate | |
Crude homogenate of normal human colorectal mucosa | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction