Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myofibroblast Marker Antibody (PR 2D3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250524B
Description
Myofibroblast Marker Monoclonal specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Myofibroblast Marker | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Crude homogenate of normal colorectal mucosa. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
PR 2D3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction