Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myofibroblast Marker Antibody (PR 2D3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Myofibroblast Marker |
---|---|
Clone | PR 2D3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Description
Myofibroblast Marker Monoclonal antibody specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
Myofibroblast Marker | |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Crude homogenate of normal colorectal mucosa. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
PR 2D3 | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title