Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Myogenin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Catalog No. MS1113PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1113PABX 200 μL Concentrated 1mg/mL
MS1113P1ABX 100 μL Concentrated 1mg/mL
MS1113R1 1 mL Prediluted Prediluted
MS1113R7 7 mL Prediluted Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1113PABX Supplier Epredia™ Supplier No. MS1113PABX
Add to Cart
Add to Cart

Deliver accurate, reproducible results with the Epredia™ Myogenin Ab-1, Mouse Monoclonal Antibody.

Myogenin is a member of a family of myogenic regulatory genes, which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors, which are essential for muscle development. Transfection of myogenin into multipotential mesodermal cells have been shown to convert the mesodermal cells to myoblasts. Expression of myogenin is restricted to cells of skeletal muscle origin. It is therefore a useful marker for tumors of the muscle lineage, being strongly expressed in alveolar rhabdomyosarcomas.

Host Species: Mouse

Clone: F5D

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat and Cat. Does not react with quail. Others not known.

Epitope: aa 138-158

Immunogen: Recombinant protein containing rat myogenin aa 30-224

Molecular Weight: 34kDa

Positive Control: Rh30 cells, Rhabdomyosarcoma or Wilm's tumors

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Myogenin Ab-1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F5D
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein containing rat myogenin aa 30-224
Quantity 200 μL
Regulatory Status RUO
Research Discipline Muscle Biology
Primary or Secondary Primary
Target Species Feline, Human, Mouse, Rat
Form Concentrated
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.