Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myogenin Antibody (671038) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6686W
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human, Mouse samples. It is validated for ImmunocytochemistrySpecifications
Myogenin | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Immunocytochemistry | |
671038 | |
Immunocytochemistry | |
BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
E. coli-derived recombinant human Myogenin, Ser124-Asn224, Accession # P15173 | |
0.1 mL | |
Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
4656 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction