Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myogenin Antibody (rMYOG/6297), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314012C
This item is not returnable.
View return policy
Description
Myogenin Monoclonal antibody specifically detects Myogenin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Myogenin | |
| Monoclonal | |
| DyLight 650 | |
| BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
| Recombinant full-length human Myogenin protein (Uniprot: P15173) | |
| 0.1 mL | |
| Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
| 4656 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| rMYOG/6297 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction