Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myogenin Mouse anti-Human, Clone: MYOG/2660, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Myogenin |
---|---|
Clone | MYOG/2660 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Myogenin Monoclonal specifically detects Myogenin in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
Myogenin | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Cancer, Stem Cell Markers, Transcription Factors and Regulators | |
10 mM PBS with 0.05% BSA | |
4656 | |
IgG1 κ | |
Protein A or G purified |
MYOG/2660 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) | |
Recombinant full-length human Myogenin (MYOG) protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title