Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myogenin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116750
Description
Positive Samples: Rat skeletal muscle.
Myogenin is a muscle-specific transcription factor that can induce myogenesis in a variety of cell types in tissue culture. It is a member of a large family of proteins related by sequence homology, the helix-loop-helix proteins. It is essential for the development of functional skeletal muscle.
Specifications
Myogenin | |
Polyclonal | |
Unconjugated | |
MYOG | |
bHLHc3; Class C basic helix-loop-helix protein 3; MYF4; Myf-4; myo; MYOD1-related protein; MYOG; Myogenic factor 4; Myogenic factor-4; myogenin; myogenin (myogenic factor 4) | |
Rabbit | |
Affinity chromatography | |
RUO | |
17928, 29148, 4656 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.76 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P12979, P15173, P20428 | |
MYOG | |
Synthetic Peptide. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction