Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myoglobin Monoclonal Antibody (6H8B5, 5A2G8, 4D7H3, 3H6A7)
GREENER_CHOICE

Catalog No. PIMA515305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515305 Supplier Invitrogen™ Supplier No. MA515305
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15305 targets Myoglobin in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15305 immunogen is purified recombinant fragment of Myoglobin expressed in E. Coli.

This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myoglobin
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6H8B5, 5A2G8, 4D7H3, 3H6A7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% sodium azide
Gene MB
Gene Accession No. P02144
Gene Alias AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB
Gene Symbols MB
Host Species Mouse
Immunogen Purified recombinant fragment of Myoglobin expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.