Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Myoglobin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106687
Description
Antibody detects endogenous levels of total Myoglobin.
This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
| Myoglobin | |
| Polyclonal | |
| Unconjugated | |
| MB | |
| AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4151 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02144 | |
| MB | |
| A synthesized peptide derived from human MB(Accession P02144), corresponding to amino acid residues E28-K78. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction