Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Myoglobin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110666 Supplier Invitrogen™ Supplier No. PA5110666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGEWQLVLNV WGKVEADIPG HGQEVLIRLF KGHPETLEKF DKFKHLKSED EMKASEDLKK HGATVLTALG GILKKKGHHE AEIKPLAQSH ATKHKIPVKY LEFISECIIQ VLQSKHPGDF GADA Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 83%.

This gene encodes a member of the globin superfamily and is expressed in skeletal and cardiac muscles. The encoded protein is a hemoprotein contributing to intracellular oxygen storage and transcellular facilitated diffusion of oxygen. At least three alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Myoglobin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MB
Gene Accession No. P02144
Gene Alias AI325109; Human myoglobin; Mb; MGC13548; myoglobgin; myoglobin; OTTHUMP00000197936; PVALB
Gene Symbols MB
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Myoglobin. Recombinant protein control fragment (Product #RP-91501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4151
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.